Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77882

Supelco

6-Hydroxyindole

analytical standard

Synonym(s):

6-Indolol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NO
CAS Number:
Molecular Weight:
133.15
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-30°C

SMILES string

Oc1ccc2cc[nH]c2c1

InChI

1S/C8H7NO/c10-7-2-1-6-3-4-9-8(6)5-7/h1-5,9-10H

InChI key

XAWPKHNOFIWWNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
  • Reactant for asymmetrical synthesis of notoamide J as a potential biosynthetic precursor of prenylated indole alkaloids
  • Reactant for preparation of (quinolinyloxymethyl)isoxazolecarboxylate esters antituberculosis agents
  • Reactant for preparation of indolyl(propanolamine) derivatives as HIV inhibitors
  • Reactant for preparation of indoleoxyacetic acid derivatives as peroxisome proliferator-activated receptor agonists
  • Reactant for preparation of 1-aroylindole 3-aroylindoles combretastatin A-4 analogs as antitumor agents and tubulin polymerization inhibitors

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service