Skip to Content
MilliporeSigma
All Photos(1)

Documents

74872

Supelco

Rutin trihydrate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Quercetin-3-rutinoside trihydrate, Vitamin P trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O16 · 3H2O
CAS Number:
Molecular Weight:
664.56
Beilstein:
75455
MDL number:
UNSPSC Code:
85151701

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

expiry date on the label
limited shelf life

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

food and beverages

storage temp.

2-8°C

SMILES string

O.O.O.C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(Oc4cc(O)cc(O)c4C3=O)c5ccc(O)c(O)c5)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H30O16.3H2O/c1-8-17(32)20(35)22(37)26(40-8)39-7-15-18(33)21(36)23(38)27(42-15)43-25-19(34)16-13(31)5-10(28)6-14(16)41-24(25)9-2-3-11(29)12(30)4-9;;;/h2-6,8,15,17-18,20-23,26-33,35-38H,7H2,1H3;3*1H2/t8-,15+,17-,18+,20+,21-,22+,23+,26+,27-;;;/m0.../s1

InChI key

NLLBWFFSGHKUSY-JPRRWYCFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Other Notes

This compound is commonly found in plants of the genus: achillea crataegus echinacea eucalyptus ginkgo glycyrrhiza hedera humulus hypericum lycium pimpinella primula sambucus serenoa

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service