Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59037

Column Frits

diam. 1/8 in., pore size 0.5 μm

Synonym(s):

0.5 μm pores

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

description

for Conventional Column Hardware (compression style)

packaging

pkg of 10 ea

diam.

1/8 in.

thickness

0.039 in.

pore size

0.5 μm

Looking for similar products? Visit Product Comparison Guide

General description

Choose frits with 0.5 μm pores to retain packings prepared from 3 μm particles, frits with 2 μm pores to retain larger particles.
Column frits is a porous frit, which is widely used for column protection. It keeps the packing material in the column.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

High Performance Liquid Chromatography:Principles And Methods In Biotechnology (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service