Key Documents
58806
Deburring Tool
configured for 1/8 in. tubing
Synonym(s):
Dressing tool
About This Item
UNSPSC Code:
41115700
packaging
pkg of 1 ea
compatibility
configured for 1/8 in. tubing
General description
Deburring tool is an accessory that can be used with 1/8 in. tubing for dressing it without distortion after being smooth-cut using the tubing cutter.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)