Key Documents
58632
Upchurch Fingertight Fittings Kit
About This Item
UNSPSC Code:
41115700
Pricing and availability is not currently available.
General description
High pressure fittings with 10-32 threads for 1/16 in. O.D. tubing. Kit includes:
  • Delrin Fingertight nut, good to 4,000 psi (280 kg/cm2), 5 ea
  • Stainless steel Fingertight nut, good to 6,000 psi (420 kg/cm2), wrenchtight to 10,000 psi (700 kg/cm2), 5 ea
  • PEEK gripping ferrule, 20 ea.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1