Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58348

Supelco

SUPELCOSIL LC-18-DB (5 µm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

SUPELCOSIL LC-18-DB HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm

Agency

suitable for USP L1

Quality Level

feature

endcapped

manufacturer/tradename

SUPELCOSIL

extent of labeling

11.0% Carbon loading

parameter

0-70 °C temperature
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

surface area

170 m2/g

surface coverage

surface coverage 3.1 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

120 Å

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

SUPELCOSIL LC-DB phases are specially deactivated for basic compounds. These columns provide shorter retention, better peak shape, and higher efficiency for organic bases than can be obtained on other Type A silica reversed-phase columns.

Application

SUPELCOSIL LC-18-DB HPLC Column has been used for the separation of:
  • Analytes of pharmaceutical relevance during their analysis using LC in conjunction with principal component analysis (PCA).
  • Loratadine in human plasma during its analysis by using HPLC combined with ultraviolet (UV) detection.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of loratadine in human plasma by high-performance liquid chromatographic method with ultraviolet detection.
Kunicki PK.
Journal of Chromatography. B, Biomedical Sciences and Applications, 755(1-2), 331-335 (2001)
Chromatographic classification and comparison of commercially available reversed-phase liquid chromatographic columns using principal component analysis
Euerby MR and Petersson P
Journal of Chromatography A, 994(1-2), 13-36 (2003)
Jaganathan Subramani et al.
Aging, 12(19), 19809-19827 (2020-10-14)
Aging is an independent risk factor for cardiovascular diseases, such as myocardial infarction due to ischemia-reperfusion injury (I/R) of the heart. Cytosolic thioredoxin (Trx) is a multifunctional redox protein which has antioxidant and protein disulfide reducing properties. We hypothesized that
E Terjéki et al.
Journal of pharmaceutical and biomedical analysis, 24(5-6), 913-920 (2001-03-15)
RGH-1756 (1-(2-methoxy-phenyl)-4-(4-[4-(6-imidazo[2,1-b]-thiazolyl)-phenoxy]-butyl)-piperazine dimethansulphonate) is a novel atypical antipsychotic candidate of Gedeon Richter Ltd. A new HPLC method has been developed and validated for the quantitative determination of RGH-1756 in dog and rat plasma. The compound and the internal standard are
J S Patrick et al.
Analytical biochemistry, 199(1), 125-131 (1991-11-15)
A rapid, isocratic method for the determination of tryptophan in Escherichia coli fermentation broths by reversed-phase HPLC is described. Tryptophan can be measured in fermentations containing either chemically defined media or media with hydrolyzed protein supplements. The procedure was rugged

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service