Skip to Content
MilliporeSigma
All Photos(2)

Documents

575650-U

Supelco

PlatePrep 96-well Vacuum Manifold

Starter kit

Synonym(s):

96-well SPE plate vacuum starter kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
NB.21

material

polypropylene tube

Quality Level

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

The PlatePrep vacuum manifold consists of a clear acrylic top allowing for easier inspection of flow rates during SPE 96-well plate processing. The polypropylene base offers excellent chemical resistance while a single remote vacuum gauge/bleed valve controls flow through all the wells.

Use this compact vacuum manifold in conjunction with a Discovery SPE 96-well plate to process up to 96 samples concurrently. The single valve control, parallel processing capabilities, and uniform flow dynamics allow for easier method development, reduce clutter, and improve reproducibility. Unused wells can be covered and used at a later date.

A PlatePrep 96-Well Starter Kit (Cat. No. 575650-U) is available to anaysts new to 96-well SPE technology. Included with the Starter Kit is PlatePrep Vacuum Manifold and sample units of key accessory items necessary for 96-well SPE.

Components

Starter Kit includes:
  • 1 PlatePrep vacuum manifold
  • 1 96 square well collection plate, 2 mL, polypropylene
  • 2 disposable reservoir/waste trays, PVC
  • 1 96 sq. well pierceable cap mat
  • 5 reagent reservoirs
  • 1 cluster tube rack

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew J Dominguez et al.
Protein expression and purification, 152, 84-91 (2018-07-25)
Protein purification is essential in the study of protein structure and function, and the development of novel therapeutics. Many studies require purifying multiple proteins at once, increasing the demand for improved purification methods. We hypothesized that multiple chromatography columns could

Articles

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

Protocols

SEC-MS protocol for rapid glycoprofiling of monoclonal antibodies with antibody purification and mass spectrometer calibration.

SEC-MS protocol for rapid glycoprofiling of monoclonal antibodies with antibody purification and mass spectrometer calibration.

SEC-MS protocol for rapid glycoprofiling of monoclonal antibodies with antibody purification and mass spectrometer calibration.

SEC-MS protocol for rapid glycoprofiling of monoclonal antibodies with antibody purification and mass spectrometer calibration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service