Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57410-U

Knitted reactor coil

L × I.D. 20 m × 0.25 mm, volume 1.0 mL

Synonym(s):

Coiled reactor bundle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
SB.51

L × I.D.

20 m × 0.25 mm

volume

1.0 mL

compatibility

for use with PHRED Reactor

General description

Knitted reactor coil is used with PHRED photochemical reactor, to determine the duration of exposure to ultraviolet radiation (UV) for the detection of analytes.

Application


  • Use of an on-line, precolumn photochemical reactor in high-performance liquid chromatography of naphthodianthrones in Hypericum perforatum preparations.: This study explores the integration of a knitted reactor coil as a precolumn photochemical reactor in HPLC, optimizing the detection of naphthodianthrones in Hypericum perforatum. The results indicate improved sensitivity and specificity in chromatographic analysis (Schmidt, 2003).

  • Optimization of post-column photolysis and electrochemical detection for the liquid chromatographic determination of 3-nitro-L-tyrosine.: This research details the optimization of a knitted reactor coil in post-column photolysis and electrochemical detection in HPLC for 3-nitro-L-tyrosine. The findings demonstrate enhanced analytical performance, highlighting the reactor′s role in increasing method sensitivity (Liu et al., 1998).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Jahn et al.
Pharmaceutical research, 37(6), 118-118 (2020-06-05)
Polysorbates are critical stabilizers in biopharmaceutical protein formulations. However, they may degrade in drug substance (DS) or drug product (DP) during storage. Degradation catalyzed by lipases present in host cell proteins (HCPs) is one suspected root cause. The purpose of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service