Skip to Content
MilliporeSigma
All Photos(1)

Documents

57109-U

Supelco

Supelclean ENVI-Carb SPE Tube

bed wt. 100 mg, volume 1 mL, pk of 108

Synonym(s):

Carbon SPE cartridge tube, 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41115712
NACRES:
NB.21

material

polypropylene tube

Quality Level

product line

Supelclean

composition

bed wt., 100 mg

packaging

pk of 108

technique(s)

solid phase extraction (SPE): suitable

surface area

100 m2/g

volume

1 mL

matrix active group

carbon phase

particle size

120-400 mesh

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Graphitized Non-Porous Carbon

Retention Mechanism: Reversed-phase
Sample Matrix Compatibility: Aqueous solutions (drinking, ground, waste water)

  • Surface area: 100m2/g; Particle Sz.:120/400 mesh
  • Extreme affinity for organic polar and non-polar compounds from both non-polar and polar matrices when used under reversed-phase conditions
  • Carbon surface comprised of hexagonal ring structures, interconnected and layered into graphitic sheets
  • Non-porous nature of the carbon phase allows for rapid processing, adsorption does not require analyte dispersion into solid phase pores
  • Independent investigators have found ENVI-Carb extremely useful for the rapid sample preparation of over 200 pesticides from various matrices including ground water, fruits and vegetable

Application


  • Achieving optimal balance: tuning electrical and optical characteristics of carbon electrodes for emerging photovoltaics: This research explores the utilization of toluene in enhancing the performance of carbon-based electrodes, demonstrating significant improvements in both electrical and optical properties for photovoltaic applications (Gul et al., 2024).

  • Thiol Coordination Softens Liquid Metal Particles To Improve On-Demand Conductivity: This study highlights the role of toluene in the synthesis of liquid metal particles, which are optimized for enhanced conductivity in flexible electronics, indicating a versatile application of toluene in advanced manufacturing technologies (Muller et al., 2024).

  • Using highly water-stable wool keratin/CsPbBr(3) nanocrystals as a portable amine-responsive fluorescent test strip for onsite visual detection of food freshness: Toluene′s properties are leveraged in the synthesis of innovative materials for food safety applications, showcasing its importance in the creation of sensitive and reliable detection systems (Sun et al., 2024).


Legal Information

ENVI-Carb is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Pretreatment with Mucinase StcE increases glycopeptide identification from mucin samples, enhancing sample preparation efficiency for glycopeptide analysis.

Pretreatment with Mucinase StcE increases glycopeptide identification from mucin samples, enhancing sample preparation efficiency for glycopeptide analysis.

Pretreatment with Mucinase StcE increases glycopeptide identification from mucin samples, enhancing sample preparation efficiency for glycopeptide analysis.

Pretreatment with Mucinase StcE increases glycopeptide identification from mucin samples, enhancing sample preparation efficiency for glycopeptide analysis.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service