Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57036

Supelco

Visiprep SPE Manifold Replacement Solvent Guide Needles

for use with standard, 12- and 24-port models (optional use with 57030-U & 57250-U), stainless steel, pk of 12

Synonym(s):

V.M.SS SOL.GUIDE NEEDLES 12/PK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

material

stainless steel

Agency

suitable for DIN 38407-42
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for GB 31604.35-2016
suitable for ISO 21675 2019
suitable for ISO 25101

reg. compliance

suitable for FDA C-010.02

packaging

pk of 12

technique(s)

solid phase extraction (SPE): suitable

application(s)

PFAS testing
food and beverages

compatibility

for use with standard, 12- and 24-port models (optional use with 57030-U & 57250-U)

Looking for similar products? Visit Product Comparison Guide

General description

Visiprep SPE Manifold Replacement Solvent Guide Needles are accessories for use with 12-port or 24-port Visiprep solid-phase extraction manifolds.

Legal Information

Visiprep is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service