57033
Visiprep™ Manifold Replacement Gasket
for use with 12-port model (standard and DL), pk of 2
Synonym(s):
V.M. GASKETS 2/PK
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
Recommended Products
packaging
pk of 2
Quality Level
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
compatibility
for use with 12-port model (standard and DL)
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Visiprep is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service