Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56908

Supelco

TraceCERT® Multiresidue Pesticide Standard Mix #8

in tert-butyl methyl ether, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

in tert-butyl methyl ether
100 μg/mL each component in tert-butyl methyl ether
100 μg/mL in tert-butyl methyl ether

application(s)

cannabis testing
food and beverages
(pesticide residue testing)

format

multi-component solution

storage temp.

−20°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Components

The mix contains the following analytes:

Endosulfan sulfate
alpha-Endosulfan
beta-Endosulfan
Etridiazole
Fenvalerate
Kinoprene-S
MGK-264

Other Notes

7 components. When combined with other mixes, suitable for use in meeting the requirements of mulitple jurisdictions for pesticide testing of cannabis products
Check out our entire range of pesticide mixes for Cannabis testing

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-18.4 °F

Flash Point(C)

-28 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct extraction with acetonitrile of hemp seed oil for the analysis of pesticides by using comprehensive two-dimensional gas chromatography-triple quadrupole mass spectrometry
Arena A, et al.
Journal of Chromatography A, 464432-464432 (2023)
Alessia Arena et al.
Journal of chromatography. A, 1710, 464432-464432 (2023-10-14)
The method herein described involves a rapid and limited-volume (0.5 mL of acetonitrile) solvent-extraction sample preparation process, for pesticide determination in hemp seed oil. The extraction method was characterized by the absence of both clean-up or pre-concentration steps. The extracts were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service