Key Documents
55075
Upchurch Tubing Adapter
About This Item
UNSPSC Code:
41115700
material
red PEEK adapter (one-piece design)
packaging
pkg of 1 ea
fitting
female Luer (lock rim)
1/4-28 female
General description
Upchurch Tubing Adapter is a one piece design used to connect membrane adsorber units to a workstation. It is made of PEEK.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)