Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

548676-U

Supelco

Smart SPME Fiber Assembly

greener alternative

PEG, for use with PAL 3, Series 2 Smart autosampler, Nitinol-core (NIT), 23 ga, 60 μm (PEG)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.21

Product Name

Smart SPME Fiber Assembly, Carbowax-Polyethylene Glycol (PEG), df 60 μm(PEG, for use with PAL 3, Series II Smart autosampler, size 23 ga

material

metal alloy fiber
purple hub

size

23 ga

packaging

pkg of 1 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

60 μm (PEG)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

PEG coating

compatibility

(for alcohols and polar compounds (MW 40 - 275))
for use with PAL 3, Series II Smart autosampler

greener alternative category

General description

Smart SPME fiber assemblies combine innovative SPME devices with Smart technology for seamless sample preparation. Each Smart fiber assembly, specifically designed to be compatible with the PAL 3, Series II Smart autosampler, incorporates a SPME fiber within a holder and a programmable computer chip coded with fiber chemistry type, dimensions, and lot number. The Smart chip provides traceability by recording fiber stroke count, injection and conditioning durations, usage dates, and expiration date. The one piece fiber/holder design makes the Smart SPME assembly easier to handle and install than traditional SPME. In addition, compatibility with the SPME tool on the PAL 3 autosampler allows for more efficient method development and completely automated sample preparation, thus increasing overall productivity.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service