Skip to Content
MilliporeSigma

Skip To

54849

SUPELCOSIL LC-18 (5 µm) HPLC Columns

L × I.D. 25 cm × 21.2 mm, HPLC Column

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
NB.21
L × i.d.:
25 cm × 21.2 mm
Particle size:
5 μm
Matrix active group:
C18 (RP18), C18 (octadecyl) phase
Pore size:
120 Å
Matrix:
silica gel, spherical particle platform
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

SUPELCOSIL LC-18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm

agency

suitable for USP L1

Quality Segment

feature

endcapped

manufacturer/tradename

SUPELCOSIL

extent of labeling

11.0% carbon loading

parameter

≤70 °C temp. limit, 400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 21.2 mm

surface area

170 m2/g

surface coverage

surface coverage 3.1 μmol/m2

matrix

silica gel, spherical particle platform

matrix active group

C18 (RP18), C18 (octadecyl) phase

particle size

5 μm

pore size

120 Å

pH

2-7.5

application(s)

food and beverages

separation technique

reversed phase

General description

A general purpose hydrophobic alkyl phase that is very retentive and gives good peak shape for a wide variety of compounds.

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
579345793558239
separation technique

reversed phase

separation technique

reversed phase

separation technique

reversed phase

separation technique

reversed phase

particle size

5 μm

particle size

5 μm

particle size

5 μm

particle size

5 μm

matrix active group

C18 (octadecyl) phase

matrix active group

C18 (octadecyl) phase

matrix active group

C18 (octadecyl) phase

matrix active group

C18 (octadecyl) phase

L × I.D.

25 cm × 21.2 mm

L × I.D.

15 cm × 2.1 mm

L × I.D.

25 cm × 2.1 mm

L × I.D.

5 cm × 4.6 mm

pore size

120 Å

pore size

120 Å

pore size

120 Å

pore size

120 Å

matrix

silica gel, spherical particle platform

matrix

silica gel, spherical particle platform

matrix

silica gel, spherical particle platform

matrix

silica gel, spherical particle platform


Still not finding the right product?

Explore all of our products under SUPELCOSIL LC-18 HPLC Column




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters