Skip to Content
MilliporeSigma

54792

LiChrospher® Si-60 (5 µm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, HPLC Column

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
NB.21
L × i.d.:
25 cm × 4.6 mm
Particle size:
5 μm
Matrix active group:
Si, silica phase
Pore size:
60 Å
Matrix:
silica gel
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

LiChrospher® Si-60 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

Quality Segment

agency

suitable for USP L3

manufacturer/tradename

LiChrospher®

parameter

≤70 °C temp. range, 400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

650 m2/g

matrix

silica gel

matrix active group

Si, silica phase

particle size

5 μm

pore size

60 Å

application(s)

food and beverages

separation technique

hydrophilic interaction (HILIC), normal phase

General description

LiChrospher® is a reliable and versatile spherical silica sorbent, which is traditionally produced and available with different modifications. LiChrospher® Si 60 (5 μm particle size, L × I.D. 25 cm × 4.6 mm) is ideal for normal-phase HPLC chromatography separations.

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under LiChrospher® Si-60 HPLC Column




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
5479204061833666975