Skip to Content
MilliporeSigma
All Photos(1)

Documents

52972-U

Supelco

Supelclean Z-Sep+ SPE Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104018
NACRES:
NB.21

material

PE frit (20/40 μm)
polypropylene hardware

composition

Bed wt., 500 mg

packaging

pk of 30 ×

technique(s)

solid phase extraction (SPE): suitable

volume

6 mL

application(s)

food and beverages

General description

This product was developed for pesticide residue analysis in livestock products. Use for samples that contain low or intermediate polarity pesticides.

Legal Information

Supelclean is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service