Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48570

Supelco

Pyrene

analytical standard

Synonym(s):

Benzo[def]phenanthrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10
CAS Number:
Molecular Weight:
202.25
Beilstein:
1307225
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

145-148 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

c1cc2ccc3cccc4ccc(c1)c2c34

InChI

1S/C16H10/c1-3-11-7-9-13-5-2-6-14-10-8-12(4-1)15(11)16(13)14/h1-10H

InChI key

BBEAQIROQSPTKN-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

Pyrene belongs to polycyclic aromatic hydrocarbon group. These are organic compounds which have two or more fused benzene rings in linear, angular and cluster arrangements. Pyrene is a four ring PAH.
Pyrene is a neat reference standard, which may be used for environmental analysis.

Application

Pyrene may be used as an analytical reference standard for the quantification of the analyte in environmental tobacco smoke samples using high-performance liquid chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

435.2 °F

Flash Point(C)

224 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Semivolatile and particulate polycyclic aromatic hydrocarbons in environmental tobacco smoke: cleanup, speciation, and emission factors
Gundel AL, et al.
Environmental Science & Technology, 259(6), 1602-1614 (1995)
Pentti Somerharju
Chemistry and physics of lipids, 116(1-2), 57-74 (2002-07-03)
Pyrene is one of the most frequently used lipid-linked fluorophores. Its most characteristic features are a long excited state lifetime and (local) concentration-dependent formation of excimers. Pyrene is also hydrophobic and thus does not significantly distort the conformation of the
Michael E Østergaard et al.
Chemical Society reviews, 40(12), 5771-5788 (2011-04-14)
Pyrene-functionalized oligonucleotides (PFOs) are increasingly explored as tools in fundamental research, diagnostics and nanotechnology. Their popularity is linked to the ability of pyrenes to function as polarity-sensitive and quenchable fluorophores, excimer-generating units, aromatic stacking moieties and nucleic acid duplex intercalators.
Intramolecular pyrene excimer fluorescence: a probe of proximity and protein conformational change.
S S Lehrer
Methods in enzymology, 278, 286-295 (1997-01-01)
Yan-Jun Huang et al.
Journal of the American Chemical Society, 135(5), 1700-1703 (2013-01-16)
Aggregates of an amphiphilic monoboronic acid bearing a hydrophobic pyrene fluorophore were employed for highly modulating, sensitive, and selective ratiometric fluorescent sensing of glucose in aqueous solution. The selectivity for glucose was improved by "knock-out" binding of fructose by phenylboronic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service