Skip to Content
MilliporeSigma
All Photos(1)

Documents

48479

Supelco

ASTM® D5307 Crude oil internal standard

certified reference material, (equal weights of the hydrocarbons listed)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Quality Level

Agency

ASTM® D5307

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 25 mL

concentration

(equal weights of the hydrocarbons listed)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

Application

ASTM® D5307 crude oil internal standard has been used in the quantitative determination of diamondoid compounds like adamantane in specific organic solvents like dichloromethane, hexane, carbon-tetrachloride, hexane , heptadecane, hexadecane, pentadecane and tetradecane, using infrared attenuated total reflection spectroscopy (IR-ATR).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Equal weights of the hydrocarbons listed. Add to the crude oil sample as the internal standard.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Heptadecane 25 % (w/w)
    Hexadecane 25 % (w/w)
    Pentadecane 25 % (w/w)
    Tetradecane 25 % (w/w)

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of adamantane in organic media via infrared attenuated total reflection spectroscopy
Luzinova Y, et al.
Organic Geochemistry, 40, 1143-1150 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service