Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48146

Supelco

N,N′-1,6-Hexanediylbis(4-(2-pyridinyl)-1piperazinecarboxamide) solution (derivative of 1,6-HDIP)

certified reference material

Synonym(s):

N,N′-1,6-Hexanediylbis{4-(2-pyridinyl)-1-piperazinecarboxamide} solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

ASTM® D5836
OSHA 5002

description

derivative of 1,6-HDIP

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

SMILES string

N2(CCN(CC2)C(=O)Nc3c(cccc3)Cc4c(cccc4)NC(=O)N5CCN(CC5)c6ncccc6)c1ncccc1

InChI

1S/C33H36N8O2/c42-32(40-21-17-38(18-22-40)30-13-5-7-15-34-30)36-28-11-3-1-9-26(28)25-27-10-2-4-12-29(27)37-33(43)41-23-19-39(20-24-41)31-14-6-8-16-35-31/h1-16H,17-25H2,(H,36,42)(H,37,43)

InChI key

MZYVHJRNOMUNAH-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
ASTM is a registered trademark of American Society for Testing and Materials

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service