Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47427-U

Supelco

EPA 524.2 Rev 4 Update Mix

2000 μg/mL in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 524.1,524.2

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

EPA 524.2 Rev 4 Update Mix may be utilized for identifying and quantifying volatile organic compounds (VOCs) in drinking water. The pre-requisite of EPA 524.2 method is the use of gas chromatography with a selective detector, gas chromatography/mass spectrometry or high performance liquid chromatography.

Application

EPA 524.2 Rev 4 Update Mix may be used as an analytical standard to quantify volatile organic compound (VOC) analytes in indoor, outdoor, and occupational environments by gas chromatography coupled to mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Acrylonitrile
    Allyl chloride
    tert-Butyl methyl ether
    Carbon disulfide
    Chloroacetonitrile
    1-Chlorobutane
    trans-1,4-Dichloro-2-butene
    Diethyl ether
    Ethyl methacrylate
    Hexachloroethane
    Methacrylonitrile
    Methyl acrylate
    Methyl methacrylate
    Nitrobenzene
    2-Nitropropane
    Pentachloroethane
    Propionitrile
    Tetrahydrofuran
    See All (18)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diffusive uptake rates of volatile organic compounds on standard ATD tubes for environmental and workplace applications
Jia C and Fu X
Chemosphere, 4(4), 87-87 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service