Skip to Content
MilliporeSigma
All Photos(1)

Documents

47084-U

Supelco

Amide Test Mixture

in acetonitrile: water (1:1) (varied conc.), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

packaging

ampule of 1 mL

concentration

in acetonitrile: water (1:1) (varied conc.)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

compatibility

use to QA hydrophobic reversed phase columns

format

multi-component solution

storage temp.

room temp

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Analyte

Description

    Uracil 7 μg/L
    Acetophenone 7 μg/L
    Benzene 750 μg/L
    Toluene 775 μg/L

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service