Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28397-U

Supelco

Multi-Layer Silica Gel Dioxin Column

for use with Dioxin Sample Preparation Kit, O.D. × L 6.35 mm × 35 cm

Synonym(s):

Glass Silica Gel Column (for EPA 1631)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NB.21

material

glass hardware

Quality Level

Agency

suitable for EPA 1613

packaging

pkg of 5

technique(s)

solid phase extraction (SPE): suitable

I.D. × L

15 mm × 35 cm

O.D. × L

6.35 mm × 35 cm

thread I.D.

24 mm

application(s)

food and beverages

compatibility

for use with 6.35 mm unions
for use with Dioxin Sample Preparation Kit

General description

Multi-Layer Silica Gel Dioxin Column is compatible with the Dioxin Sample Preparation Kit. It is suitable for the solid phase extraction (SPE) technique.

It can be used for the purification of extracts containing polychlorinated dibenzo-p-dioxins (PCDDs), polychlorinated dibenzofurans (PCDFs), and polychlorinated biphenyls (PCBs), helping to remove impurities and isolate these target compounds for analysis.

Application

The Multi-Layer Silica Gel Dioxin Column has been used as a cleanup process to remove potential chromatographic interferences and prepare the samples for further analysis and quantification during the USEPA method.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioaccumulation of dioxins in the benthic fish from Hiroshima Bay
Hiroyuki T, et al.
Environmental Chemistry, 2, 47-54 (2009)
Developing an improved strategy for the analysis of polychlorinated dibenzo-p-dioxins/furans and dioxin-like polychlorinated biphenyls in contaminated soils using a combination of a one-step cleanup method and gas chromatography with triple quadrupole mass spectrometry.
Haena C, et al.
Toxics, 11, 738-738 (2023)

Related Content

Solid phase extraction purifies samples for chromatographic analysis, improving data quality and protecting analytical columns.

Solid phase extraction purifies samples for chromatographic analysis, improving data quality and protecting analytical columns.

Solid phase extraction purifies samples for chromatographic analysis, improving data quality and protecting analytical columns.

Solid phase extraction purifies samples for chromatographic analysis, improving data quality and protecting analytical columns.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service