Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28034-U

Supelco

Capillary Column Nut (fits Agilent [MS detector] and PerkinElmer®), hexagonal wrenchtight version

pkg of 5 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
SB.54

General description

These nickel-plated brass column nuts are replacements for damaged or misplaced original Agilent MSD or PerkinElmer nuts. Use with long ferrules.

Legal Information

PerkinElmer is a registered trademark of PerkinElmer Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

2-layer SPE cartridge efficiently reduces background for ginger contaminant analysis.

2-layer SPE cartridge efficiently reduces background for ginger contaminant analysis.

2-layer SPE cartridge efficiently reduces background for ginger contaminant analysis.

2-layer SPE cartridge efficiently reduces background for ginger contaminant analysis.

See All

Protocols

Paprika spice's PAH contamination risk due to environmental exposure highlights food safety concerns.

Paprika spice's PAH contamination risk due to environmental exposure highlights food safety concerns.

Paprika spice's PAH contamination risk due to environmental exposure highlights food safety concerns.

Paprika spice's PAH contamination risk due to environmental exposure highlights food safety concerns.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service