Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27748-U

domnick hunter® NITROX UHP Hydrogen Generator

Model 20H, output flow, 0 ‑ 160 cc/min

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
NACRES:
SB.54

feature

output flow 0 ‑ 160 cc/min

manufacturer/tradename

Domnick Hunter 20H

Looking for similar products? Visit Product Comparison Guide

General description

domnick hunter hydrogen generators are a safe and economic alternative to high pressure gas cylinders. These generators deliver a continuous stream of 99.999% pure hydrogen, suitable for use with any laboratory instrument requiring a supply of ultra high purity hydrogen, and ideal for carrier gas and fuel gas applications.

domnick hunter hydrogen generators use a proprietary ion exchange membrane to produce ultra-high-purity hydrogen from laboratory grade deionized water, totally eliminating the need for caustic solution.

These generators are designed for total safety and highly reliable operation. Hydrogen is generated only on demand, at low pressure, and minimal volume is stored within the unit. The unique digital display gives system status and diagnostic capabilities not available on any other competitive hydrogen generator.

CE Approved.

Legal Information

NITROX is a trademark of domnick hunter group PLC
domnick hunter is a registered trademark of Parker Hannifin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service