Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27600-U

Supelco

Supelco® Helium Purifier

stainless steel fittings, 1/8 in. O.D. Swagelok (nuts and ferrules included)

Synonym(s):

Supelco® HELIUM GAS PURIFIER, 1/8IN FITTINGS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.54

material

stainless steel fittings

Quality Level

fitting

1/8 in. O.D. Swagelok (nuts and ferrules included)

Looking for similar products? Visit Product Comparison Guide

General description

This multiple bed trap removes hydrocarbons, moisture, oxygen, carbon monoxide, and carbon dioxide from helium gas streams. The amount of highly effective, high capacity adsorbent material in each bed is optimized so that breakthrough of each contaminant is as close to simultaneous as possible. This avoids the costly replacement observed with other broad-spectrum traps that have exhausted their capacity for one contaminant, but still have capacity for other contaminants. This trap will easily purify 99.997% purity helium to a cumulative level of 100 ppb (hydrocarbons + moisture + oxygen + carbon monoxide + carbon dioxide).

Specifications:
  • Output Purity: 99.99999%
  • Output Efficiency (total): <100 ppb
  • Output Efficiency (hydrocarbons): <20 ppb (as methane)
  • Output Efficiency (moisture): <10 ppb
  • Output Efficiency (oxygen): <2 ppb
  • Output Efficiency (carbon monoxide): <20 ppb
  • Output Efficiency (carbon dioxide): <1ppm
  • Capacity (hydrocarbons): 30 g (as methane)
  • Capacity (moisture): 46 g
  • Capacity (oxygen): 1200mg
  • Max. Flow Rate: 8 L/min.
  • Max. Pressure: 500 psi
  • Max. Temp.: 100 °C

Dimensions for both purifiers
16.5 in. (41..91 cm) long × 2 5/16 in. (6.3 cm) diameter

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorge E Spangenberg et al.
Analytical and bioanalytical chemistry, 411(10), 2031-2043 (2019-02-05)
Recently, we reported that the carbon isotope composition of the solid residues obtained by freeze-drying white and red wines (δ13CWSR) could be used for tracing the water status of the vines whose grapes were used to produce them. Here, we

Articles

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

See All

Related Content

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service