Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

26702

Hamilton® syringe, 600 series

62RNR, volume 2.5 μL, needle size 22s ga, needle L 51 mm (2 in.)

Synonym(s):

Hamilton® Syringe 62RNR, 2.5UL 22S GA, 2IN, PT#3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

blunt tip stainless steel needle (point style 3)

needle size

22s ga

description

Microliter

feature

removable needle

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 87942

needle L

51 mm (2 in.)

volume

2.5 μL

compatibility

for use with Rheodyne

Looking for similar products? Visit Product Comparison Guide

General description

Hamilton® syringe is available in gas-tight type that minimizes the gas exchange in analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.

Application

It was used in microinjection system assembly to load cell culture scaffold.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.
Microliter is a trademark of Hamilton Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, fabrication and implementation of a novel multi-parameter control microfluidic platform for three-dimensional cell culture and real-time imaging.
Vickerman, Vernella, et al.
Lab on a chip, 8.9, 1468-1477 (2008)
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 192 -192 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service