Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

25064-U

Supelco

Male Luer Coupler

pkg of 20 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NB.24

packaging

pkg of 20 ea

technique(s)

active air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with male Luer to male Luer

General description

Male Luer Coupler is an accessory used with TDS3 TD Storage Containers. It is specifically employed with Supelclean ENVI-Carb Plus SPE Tube and SPE vacuum manifold when they are used in conjunction for the solid-phase extraction of highly polar compounds from aqueous samples.

Legal Information

ENVI-Carb is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service