Skip to Content
MilliporeSigma
All Photos(1)

Documents

24110-U

Supelco

SP®-2380 Capillary GC Column

L × I.D. 30 m × 0.25 mm, df 0.20 μm

Synonym(s):

GC column, biscyanopropyl siloxane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
eCl@ss:
32119290
NACRES:
SB.54

material

fused silica

Quality Level

Agency

meets requirements for USP G48

parameter

≤25-275 °C temperature (isothermal or programmed)

Beta value

313

df

0.20 μm

technique(s)

gas chromatography (GC): suitable

L × I.D.

30 m × 0.25 mm

matrix active group

Stabilized; poly(90% biscyanopropyl/10% cyanopropylphenyl siloxane) phase

application(s)

agriculture
chemicals and industrial polymers
clinical
food and beverages

column type

capillary highly polar

Looking for similar products? Visit Product Comparison Guide

General description

Application: A highly polar cyanosiloxane column commonly used for separation of geometric (cis/trans) fatty acid methyl ester (FAME) isomers as a group. Also useful when a highly polar general purpose column with good thermal stability is required.
USP Code: This column meets USP G48 requirements.
Phase:
  • Stabilized
  • Poly(90% biscyanopropyl/10% cyanopropylphenyl siloxane)
Temp. Limits:
  • Subambient to 275 °C (isothermal or programmed)

Application

  • SP®-2380 Capillary Column may be suitable for determination of fatty acid composition in crude oil using capillary gas chromatography, used along with normal and reverse-phase high-performance liquid chromatography (HPLC) in order to study the content and composition of tocopherols, sterols, and phospholipids in soybean oils derived from genetically-modified soybeans.
  • It was also found suitable to be used for the identification of fatty acid methyl esters from mammalian samples by fast gas chromatography.
  • Furthermore, it is also suitable for determining the hydrogenation level and the level of isomerisation development during the hydrogenation process of edible oils by capillary gas chromatography.
  • Above all that it is also suitable to be used in gas chromatography equipped with flame ionization detector (FID) for analysing the cytotoxicity of fatty acids from seed oils containing conjugated linolenic acids (CLN) on mouse tumor and human monocytic leukemia cells.

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Legal Information

SP is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosa Marchetti et al.
Applied microbiology and biotechnology, 103(19), 8267-8279 (2019-08-30)
Recycling of waste cooking oil greatly reduces the environmental impact of its disposal. As fats can give rise to high methane yields, the use of waste cooking oil for biogas production seems a promising solution. The aim of this work
Effect of genetic modification on the content and composition of bioactive constituents in soybean oil.
Mounts TL
Journal of the American Oil Chemists' Society, 73 (5), 581-586 (1996)
R Suzuki et al.
Lipids, 36(5), 477-482 (2001-07-04)
The cytotoxicity of fatty acids from seed oils containing conjugated linolenic acids (CLN) was studied. Fatty acids from pomegranate, tung, and catalpa were cytotoxic to human monocytic leukemia cells at concentrations exceeding 5 microM for pomegranate and tung and 10
Fast gas chromatography for the identification of fatty acid methyl esters from mammalian samples.
Stark K and Salem N.
Liposome Technology, 17 (8), 181-181 (2005)
E M Bondia et al.
Journal of chromatography. B, Biomedical applications, 658(2), 369-374 (1994-08-19)
Total fatty acids in plasma of neonates have been analysed as their methyl esters by gas chromatography. They were separated on a capillary column coated with a SP-2380 stationary phase. As little as 100 microliters of plasma is used for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service