Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23862-U

Supelco

Supeltex M-1 Packed Column Ferrule, 1/16 in. Column O.D.

I.D. 1/16 in., 1/16 in. O.D. Column, pkg of 200 ×  ea

Synonym(s):

GC column ferrule 1/16 in. (200 pk)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
SB.54

packaging

pkg of 200 ×  ea

I.D.

1/16 in.

compatibility

1/16 in. O.D. Column

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Characteristics: Ideal for connection to mass spectrometers. High reusability. Isothermal use only.

Material Composition: Ceramic-filled PTFE

Maximum Temperature:
250 °C

Application

Supeltex M-1 Ferrule maybe used for making connection between GC sample lines and the tube during solid adsorbent preconcentration and gas chromatographic analysis of sulfur gases.

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid adsorbent preconcentration and gas chromatographic analysis of sulfur gases.
Black MS
Analytical Chemistry, 50 (7), 848-851 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service