Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23820-U

Supelco

Pin Vise Drill Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.54

General description

Drill the exact bore you need in hard or soft ferrules. Includes pin vise and 14 drill bits:

0.33 mm (0.0135 in.)
0.40 mm (0.016 in.)
0.56 mm (0.022 in.)
0.63 mm (0.025 in.)
0.72 mm (0.028 in.)
0.77 mm (0.031 in.)
0.83 mm (0.033 in.)
0.91 mm (0.036 in.)
0.97 mm (0.038 in.)
1.02 mm (0.040 in.)
1.06 mm (0.042 in.)
1.17 mm (0.046 in.)
1.40 mm (0.055 in.)
1.61 mm (0.061 in.)

The vise handle holds all bits to keep them at your fingertips. The vise also is handy for gripping fine wire when cleaning FID jets, syringe needles, or any other small orifice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service