Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23319-U

Supelco

Gold-Plated Inlet Seal (Straight Design)

pkg of 10 ea

Synonym(s):

GC inlet seal, gold-plated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
SB.54

material

stainless steel (gold-plated)

packaging

pkg of 10 ea

Looking for similar products? Visit Product Comparison Guide

General description

The inlet seal on an Agilent GC must be regularly changed to prevent sample adsorption due to accumulation of sample residue and/or septum fragments. Supelco manufactures replacement inlet seals using a high quality stainless steel plated with pure gold to insure inertness. No brighteners are used in the plating process. This straight design seal is suitable for applications with low split flows (<200 mL/min.). Each pack includes one washer for each seal.

Application

Gold-plated inlet seal may be used in the GC injection port for thermal desorption-gas chromatography/mass spectrometry (TD-GC/MS), where residual accumulation was noticed resulting in analyte adsorption and elevating signal background. Hence, replacing the gold-plated seal in regular intervals decreased the interference.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Precautions for in-injection port thermal desorption-gas chromatography/mass spectrometry (TD-GC/MS) as applied to aerosol filter samples.
Ho, Steven Sai Hang, et al.
Atmospheric Environment, 45, 1491-1496 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service