Key Documents
22375
Chart Paper for Recorder
Blank for Spectra Physics 4270, 4290 and Varian Vista 401/402, box of 25
About This Item
UNSPSC Code:
14111500
packaging
box of 25
W × L
9.3 in. × 170 ft
Application
Analog signal from the chromatograph may be connected to a recorder with chart paper speed of 5 min/in. during an analysis to measure Glutethimide concentrations in the serum and cerebrospinal fluid (CSF) using gas chromatography.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
A.V.Bakshi U.A.Bakshi
Electronic Measurements & Instrumentation (2008)
Comparison of glutethimide concentration in the serum and cerebrospinal fluid of humans in drug overdose.
M Gold et al.
Clinical chemistry, 19(10), 1158-1161 (1973-10-01)
John Kenkel
Analytical Chemistry for Technicians , 366-366 (1994)