Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20618

Supelco

Molecular Sieve 5A Moisture Trap

volume 200 cc, 1/4 in. fitting

Synonym(s):

DRYING TUBE 1/4 INCH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

volume

200 cc

fitting

1/4 in. fitting

Looking for similar products? Visit Product Comparison Guide

General description

Moisture trap or drying tube is suitable to maintain anhydrous conditions within the apparatus. There are two types of drying tubes. It is usually prepared by placing a small, loose plug of glass wool or cotton into the constriction at the end of the tube. A drying agent is poured on top the plug. Molecular sieve 5A has the affinity for water and easily displaces other trapped gases.
Molecular Sieve 5A efficiently removes moisture and heavy hydrocarbons from compressed air, electrolytically produced hydrogen, house nitrogen, or other gases with high moisture or hydrocarbon content.

Dimensions:
  • 200 cc Trap: 26 1/4 in. (67 cm) long × 1 in. (2.5 cm) diameter
  • 750 cc Trap: 18 in. (45.7 cm) long × 2 3/8 in. (6 cm) diameter
  • S-Trap: 7 1/2 in. (19 cm) long x 5 9/16 in. (14.1 cm) wide x 1/2 in. (13 mm) diameter [total bed length: 19 3/4 in. (50.2 cm); the extended bed length ensures prolonged contact between the gas and the adsorbent and provides greater working capacity]

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of atmospheric nitrous oxide using a molecular sieve 5A trap and gas chromatography.
LaHue, Miles D., Herman D. Axelrod, and James P. Lodge Jr.
Analytical Chemistry, 43, 1113-1115 (1971)
Donald Pavia
A Small Scale Approach to Organic Laboratory Techniques, 617-617 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service