Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20470

In-Line Pressure Gauge

pressure range 0-60 psi

Synonym(s):

PRESSURE GAUGE WITH TEE 0-60PSIG, EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500

parameter

0-60 psi pressure range

technique(s)

gas chromatography (GC): suitable

Looking for similar products? Visit Product Comparison Guide

General description

Install a pressure gauge in the gas delivery system to measure pressure at any given point. For example, install one upstream and downstream of the High Capacity Gas Purifier. When the pressure drop across the purifier exceeds 10 psi, the converter tube should be replaced. Contains:
  • 2 in. (5 cm) diameter steel and copper alloy gauge
  • NPT to Swagelok adapter
  • 1/8 in. tee
Nuts and ferrules included.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Balakrishnan Priya et al.
Saline systems, 6, 6-6 (2010-06-08)
Cyanobacteria are recognized as the primordial organisms to grace the earth with molecular oxygen ~3.5 billion years ago as a result of their oxygenic photosynthesis. This laid a selection pressure for the evolution of antioxidative defense mechanisms to alleviate the
James R Carter et al.
Virology journal, 11, 111-111 (2014-06-15)
Approximately 100 million confirmed infections and 20,000 deaths are caused by Dengue virus (DENV) outbreaks annually. Global warming and rapid dispersal have resulted in DENV epidemics in formally non-endemic regions. Currently no consistently effective preventive measures for DENV exist, prompting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service