Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

ZRT1374

Sigma-Aldrich

Anti-TIM-3 Antibody, clone RMT3-23 ZooMAb® Rat Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-DRP1 Antibody, clone 1E8 ZooMAb® Rabbit Monoclonal, CD366, HAVcr-2, Hepatitis A virus cellular receptor 2 homolog, T-cell immunoglobulin and mucin domain-containing protein 3, T-cell immunoglobulin mucin receptor 3, T-cell membrane protein 3, TIMD-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rat

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

RMT3-23, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 30.93 kDa
observed mol wt ~26 kDa

purified by

using protein G

species reactivity

human, rat

species reactivity (predicted by homology)

mouse

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

mouse ... Havcr2(171285)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone RMT3-23 is a ZooMAb® rat recombinant monoclonal antibody that specifically detects TIM-3.

Immunogen

Recombinant fragment corresponding to the first 191 amino acids from of Mouse TIM-3 conjugated to the Fc portion of mouse IgG2a.

Application

Quality Control Testing

Evaluated by Western Blotting in Rat thymus tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected TIM-3 in Rat thymus tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected TIM-3 in Human thymus tissue lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TIM-3 in L5178Y cells transfected with TIM-3.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected TIM-3 in one million L5178Y cells transfected with TIM-3.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Hepatitis A virus cellular receptor 2 homolog (UniProt: Q8VIM0; also known as HAVcr-2, T-cell immunoglobulin and mucin domain-containing protein 3, TIMD-3, T-cell immunoglobulin mucin receptor 3, TIM-3, T-cell membrane protein 3, CD366) is encoded by the Havcr2 (also known as Tim3, Timd3) gene (Gene ID: 171285) in murine species. TIM-3 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off to produce the mature protein that contains an extracellular domain (aa 20-193), a transmembrane domain (aa 194-214), and a cytoplasmic domain (aa 215-281). Its expression is observed in T-helper type 1 cells, but not in naïve T cells. However, its expression is up-regulated as they differentiate into T helper type 1 cells. It is also expressed by differentiated type 1 CD8+ cytotoxic T-cells, peritoneal exudate macrophages, monocytes, and splenic dendritic cells. It serves as a receptor for galectin-9 (LGALS9), phosphatidylserine, HMGB1, and CEACAM1. Binding of LGALS9 is proposed to be involved in innate immune response to intracellular pathogens. Phosphorylation of TIM-3 on tyrosine 256 is reported to increase by binding to LGALS9. Phosphorylation on tyrosine 256 and 263 also couples it to TCR signaling pathways. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Sabatos-Peyton, CA., et al. (2018). Oncoimmunology. 7(2); e1385690; Gorman, JV., et al. (2014). J. Immunol. 192(7); 3133-3142; Lee, J., et al. (2011). Mol. Cell Biol. 31(19); 3963-3974; Jayaraman, P., et al. (2010). J. Exp. Med. 207(11)l 2343-2354).

Physical form

Purified recombinant rat monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service