Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

ZRB2790

Sigma-Aldrich

Anti-p-Upf1 (Ser1127) Antibody, clone 1C4-H1 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

ATP-dependent helicase RENT1, EENT1, NORF1, Nonsense mRNA reducing factor 1, Regulator of nonsense transcripts 1, Up-frameshift suppressor 1 homolog, hUpf1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 1C4-H1
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 124.3 kDa
observed mol wt ~150 kDa

purified by

using Protein A

species reactivity

mouse, human

species reactivity (predicted by homology)

rat

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... UPF1(5976)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C4-H1 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Up-frameshift suppressor 1 homolog (Upf1) phosphorylated on serine 1127.

Immunogen

KLH-conjugated linear peptide corresponding to 13 amino acids surrounding phosphoserine 1127 in human Up-frameshift suppressor 1 homolog (Upf1).

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from A431 cells treated with Calyculin A and Okadaic Acid.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected phospho-Upf1(Ser1127) in lysate from overnight starved A431 cells treated with Calyculin A (50 nM) and Okadaic acid (500 nM) for 30 minutes, but not in lysate from untreated cells.

Tested Applications

Peptide Inhibition Assay: Target band detection in a lysate from NIH3T3 cells treated with Calyculin A (50 nM) and Okadaic acid (500 nM) was prevented by preblocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Affinity Binding Assay A representative lot of this antibody bound p-Upf1 (Ser1127) peptide with a KD of 5.6 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-Upf1 (Ser1127) in A431cells serum starved overnight then treated with Calyculin A (50 nM) and Okadaic Acid (500 nM) for 30 min.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Regulator of nonsense transcripts 1 (UniProt: Q92900; also known as ATP-dependent helicase RENT1, Nonsense mRNA reducing factor 1, NORF1, Up-frameshift suppressor 1 homolog, hUpf1) is encoded by the UPF1 (also known as KIAA0221, RENT1) gene (Gene ID: 5976) in human. Upf1 is a member of the DNA2/NAM7 helicase family. It is an RNA-dependent helicase and ATPase that is required for nonsense-mediated decay (NMD) of mRNAs containing premature stop codons. It is recruited to mRNAs upon translation termination and undergoes a cycle of phosphorylation and dephosphorylation. It is phosphorylated by SMG1 and phosphorylation is considered as an essential step in NMD and is required for formation of mRNA surveillance complexes. Phosphorylated Upf1 is recognized by EST1B/SMG5, SMG6, and SMG7, which provide a link to the mRNA degradation machinery involving exonucleolytic and endonucleolytic pathways. A hyper-phosphorylated form is targeted to the P-body, while unphosphorylated protein is distributed throughout the cytoplasm. The ATPase activity of Upf1 is required for disassembly of mRNPs undergoing NMD and is also essential for embryonic viability. Two isoforms of Upf1 have been reported that are produced by alternative splicing. Upf1 contains a UPF1-type zinc-finger domain (aa 121-272) and a nucleotide-binding domain (aa 506-510). Two isoforms of UPF1 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonal (Ref.: Chiu, S-Y., et al. (2003). RNA. 9(1); 77-87; Yamashita, A., et al. (2001). Genes Dev. 15(17); 2215-2218).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service