Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

ZRB2688

Sigma-Aldrich

Anti-Adiponectin Antibody, clone 1D19 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-Mucin-2 Antibody, clone 2C8 ZooMAb® Rabbit Monoclonal, 30 kDa adipocyte complement-related protein, ACRP30, Adipocyte C1q and collagen domain-containing protein, Adipocyte complement-related 30 kDa protein, Adipose most abundant gene transcript 1 protein, Gelatin-binding protein, apM-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D19, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 26.41 kDa
observed mol wt ~31 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... ADIPOQ(9370)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1D19 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Adiponectin. It targets an epitope within 17 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal region of human Adiponectin.

Application

Quality Control Testing

Evaluated by Western Botting in Human kidney tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Adiponectin Human kidney tissue lysate.

Tested Applications

Immunohistochemistry (Paraffin) Analysis (IHC(P)): A 1:1000 dilution of this antibody detected Adiponectin in Human kidney tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound Adiponectin peptide with a KD value of 3.7 x 10-7 in an affinity binding assay.

Immunofluorescence Analysis (IF): A 1:1000 dilution of this antibody detected Adiponectin in Human kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Adiponectin (UniProt Q15848; also known as 30 kDa adipocyte complement-related protein, ACRP30, Adipocyte complement-related 30 kDa protein, Adipocyte, C1q and collagen domain-containing protein, Adipose most abundant gene transcript 1 protein, apM-1, Gelatin-binding protein) is encoded by the ADIPOQ (also known as ACDC, ACRP30, ADPND, APM1, GBP28) gene (Gene ID: 9370) in human. Adiponectin is a homomultimeric secreted glycoprotein that is synthesized by adipocytes with a signal peptide (aa 1-18), which is subsequently cleaved off to produce the mature protein. It plays a role in the control of fat metabolism and insulin sensitivity and has anti-diabetic, anti-atherogenic, and anti-inflammatory activities. It stimulates AMPK phosphorylation and activation in the liver and the skeletal muscle and enhances glucose utilization and fatty-acid combustion. It is reported to antagonize TNF- by negatively regulating its expression in various tissues. It is also reported to inhibit NF- B signa

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service