Skip to Content
MilliporeSigma
All Photos(13)

Key Documents

ZRB2457

Sigma-Aldrich

Anti-CAM-cysteine Antibody, clone 52H11 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Carbamidomethyl-cysteine;CAM-cys

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

52H11, recombinant monoclonal

description

52H11 Clone

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

carbamidomethylation (cam) (Cys)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 52H11 is a ZooMAb® rabbit recombinant monoclonal antibody that detects Carbamidomethyl-cysteine (CAM-Cysteine) modified proteins.

Immunogen

A liner peptide corresponding to 26 amino acids from the C-terminal region of human Myeloid associated differentiation marker like 2 ( MYADML2 ) with CAM modification at two Cysteine residues.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HEK293 cells treated with Iodoacetamide.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected CAM-cysteine modified proteins in lysate from HEK293 cells treated with Iodoacetamide (1 mM for 10 min), but not in untreated cells.

Tested applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CAM-cysteine modified proteins in HEK293 cells treated with Iodoacetamide (IAM).

Enzyme Immunoassay Analysis (ELISA): A serial of dilution from a representative lot detected CAM-cysteine modified proteins in HEK293 cells treated with Iodoacetamide (IAM).

Flow Cytometry Analysis: 0.1 μg from a representative lot detected CAM-cysteine modified proteins in HEK293 cells treated with Iodoacetamide (IAM).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in lysate from HEK293 cells treated with Iodoacetamide.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected CAM-cysteine modified proteins in lysate from HEK293 cells treated with Iodoacetamide (1 mM for 10 min), but not in untreated cells.

Target description

Post-translation regulation of cysteine sulfhydryl groups can affect molecular function of proteins. In several secreted proteins, cysteine thiols undergo oxidation to form intramolecular disulfide bonds, which play key roles in protein secondary structure and function. In order to be quantified in a protein cysteine residue requires modification. Thiol group of cysteine can be blocked by a variety of reagents, including iodoacetic acid and iodoacetamide. Iodoacetate treatment can produce S-carboxymethyl derivative of cysteine, but it introduces a new negative charge into protein. However, when such charge difference is undesirable, iodoacetamide is used to derivatize cysteine to produce carboxamidomethyl cysteine, an irreversible covalent product. These modifications in proteins can serve as markers for cysteine-AGE formation and can be used as marker for mitochondrial damage during aging. Antibodies that react with carbamidomethyl-cysteine (CAM-cys) can provide multiple protein bands when subjected to Western blotting of lysates treated with iodoacetamide. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Cartee, NMP., and Wang, MM. (2020). PLoS ONE 15(9); e0239464; Cartee, NMP., et al. (2020). PLoS One. 15(11); e0242376).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service