Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

ZRB2454

Sigma-Aldrich

Anti-Ly-6C1 Antibody, clone 2H5 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-TGF β receptor 1 Antibody, clone 4H18 ZooMAb® Rabbit Monoclonal, Ly-6C.1, Lymphocyte antigen 6C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2H5, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 14.19 kDa

purified by

using Protein A

species reactivity

mouse

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

mouse ... Ly6c1(17067)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2H5 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Lymphocyte antigen 6C (Ly-6C1). It targets an epitope within 15 amino acids from the internal region.

Immunogen

Clone 2H5 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Lymphocyte antigen 6C (Ly-6C1). It targets an epitope within 15 amino acids from the internal region.

Application

Quality Control Testing

Evaluated by Western Blotting in C2C12 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Ly-6C1 in C2C12 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Ly-6C1 in NIH3T3 cell lysates.

Flow Cytometry Analysis: 1 µg from a representative lot detected Ly-6C1 in one million C57BL6 Mouse splenocytes.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Ly-6C1 in C2C12 cells.

Affinity Binding Assay: A representative lot of this antibody bound Ly-6C1 peptide with a KD of 5.2 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Ly-6C1 in Mouse spleen tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Lymphocyte antigen 6C (UniProt: P0CW02; also known as Ly-6C, Ly-6C.1) is encoded by the Ly6c1 gene (Gene ID: 17067) in murine species. Ly-6 molecules are encoded by multiple tightly linked genes located on mouse chromosome 15 and have conserved genomic organization. They are expressed on fully differentiated cells in specific compartments of peripheral lymphoid tissues. Ly-6 proteins are widely used as differentiation markers on hematopoietic cells. LY-6C is a GPI-anchor protein that is synthesized with a signal peptide (aa 1-26) and a propeptide (aa 110-131) that are subsequently cleaved off to produce the mature form (aa 27-109). It contains one UPAR/Ly6 domain that is localized in amino acids 27-115. Its expression is observed in monocytes/macrophages, endothelial cells, bone marrow cells, neutrophils, and some subsets of T cells. Its expression is considered as a superior marker to identify monocyte and macrophage subpopulations. Ly6C+ macrophages are believed to be derived from

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service