Skip to Content
MilliporeSigma
All Photos(10)

Key Documents

ZRB2058

Sigma-Aldrich

Anti-XRCC5/Ku80 Antibody, clone 1C9 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

X-ray repair cross-complementing protein 5;86 kDa subunit of Ku antigen;ATP-dependent DNA helicase 2 subunit 2;ATP-dependent DNA helicase II 80 kDa subunit;CTC box-binding factor 85 kDa subunit;CTC85;CTCBF;DNA repair protein XRCC5;Ku80;Ku86;Lupus Ku autoa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C9, recombinant monoclonal

description

1C9 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 82.71 kDa
observed mol wt ~80 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... XRCC5(7520)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C9 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects X-ray repair cross-complementing protein 5 (XRCC5). It targets an epitope within 15 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human X-ray repair cross-complementing protein 5 (XRCC5/Ku80).

Application

Quality Control Testing

Evaluated by Western Blotting in U2OS cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected XRCC/Ku80 in U2OS cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected XRCC/Ku80 in A549 cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected XRCC/Ku80 in human testis tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected XRCC/Ku80 in U2OS cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

X-ray repair cross-complementing protein 5 (UniProt: P13010; also known as 86 kDa subunit of Ku antigen, ATP-dependent DNA helicase 2 subunit 2, ATP-dependent DNA helicase II 80 kDa subunit, CTC box-binding factor 85 kDa subunit, CTC85, CTCBF, DNA repair protein XRCC5, Ku80, Ku86, Lupus Ku autoantigen protein p86, Nuclear factor IV, Thyroid-lupus autoantigen, TLAA, X-ray repair complementing defective repair in Chinese hamster cells 5) is encoded by the XRCC5 (also known as G22P2) gene (Gene ID: 7520) in human. XRCC5 is a single stranded DNA-dependent ATP-dependent helicase that plays a key role in DNA non-homologous end joining (NHEJ) by recruiting DNA-dependent protein kinase (DNA-PK) to DNA. Its Ku domain is localized in amino acids 251-460. It also contains an EEXXXDL motif (aa 720-728) that is which is required for the interaction with catalytic subunit PRKDC and its recruitment to sites of DNA damage. It exists as a heterodimer composed on XRCC5/XRCC6 (Ku80/Ku70). This dimer acts as regulatory subunit of the DNA-PK that increases the affinity of the catalytic subunit PRKDC to DNA by about 100-fold. This dimer is also suggested to act as a 5′-deoxyribose-5-phosphate lyase (5′-dRP lyase), by catalyzing the beta-elimination of the 5′ deoxyribose-5-phosphate at an abasic site near double-strand breaks. Here, XRCC5 probably acts as the catalytic subunit of 5′-dRP activity and allows to ′clean′ the termini of abasic sites. In association with N- -acetyltransferase 15 (NAA15), this dimer is reported to bind to the osteocalcin promoter and activates osteocalcin expression. In murine cells absence of either of the subunits leads to increased radiosensitivity, defective DNA double-strand break repair, and defective in V(D)J recombination. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Roberts, SA., et al. (2010). Nature. 464(7292); 1214-1217; Willis, DM., et al. (2002). J. Biol. Chem. 277(40); 37280-37291; Chan, DW., et al. (1999). Biochemistry. 38(6); 1819-1828).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

30 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service