Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

ZRB2052

Sigma-Aldrich

Anti-NFATc1 Antibody, clone 1O16 ZooMAb® Rabbit Monoclonal

enhanced validation

recombinant, expressed in HEK 293 cells

Synonym(s):

NF-ATc, NF-ATc1, NFAT transcription complex cytosolic component, NFATc, Nuclear factor of activated T-cells, cytoplasmic 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1O16, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... NFATC1(4772)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1O16 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Nuclear factor of activated T-cells, cytoplasmic 1 (NFATc1). It targets an epitope within 17 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the internal region of human Nuclear factor of activated T-cells, cytoplasmic 1 (NFATc1).

Application

Quality Control Testing

Evaluated by Western Blotting with c-Myc/DDK-tagged full-length recombinant Human NFATc1 protein.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected c-Myc/DDK-tagged, full-length, recombinant Human NFATc1 protein.

Tested Applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected NFATc1 in Jurkat cells.

Affinity Binding Assay: A representative lot of this antibody bound NFATc1 peptide with a KD of 1.0 x 10-12 in an affinity binding assay.

Enzyme Immunoassay (ELISA) Analysis: A serial of dilutions from a representative lot detected c-Myc/DKk-tagged, full-length, recombinant Human NFATc1 protein.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected NFATc1 in Human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

NuCLEAR factor of activated T-cells, cytoplasmic 1 (UniProt: O95644; also known as NF-ATc1, NFATc1, NFAT transcription complex cytosolic component, NF-ATc, NFATc) is encoded by the NFATC1 (also known as NFAT2, NFATC) gene (Gene ID: 4772) in human. NFATc1 is member of the multicomponent NFATC transcription complex that consists of at least two components, a pre-existing cytoplasmic component NFATC2 and an inducible NuCLEAR component NFATc1. NFATC proteins bind to DNA as monomers. NFATc1 that is expressed in thymus, spleen, and T-cells, plays a role in the inducible expression of cytokine genes, especially in the induction of the IL-2 or IL-4 gene transcription. NFATc1 is reported to control the cytotoxicity of mouse cytotoxic T lymphocytes. Upon primary stimulation of lymphocytes, the induction of NFATc1 occurs during the G1 phase of cell cycle. Due to an auto-regulatory feedback circuit high levels of NFATc1 are maintained during persistent immune receptor stimulation. It regulates not only the activation and proliferation, but also the differentiation and programmed death of T-lymphocytes as well as lymphoid and non-lymphoid cells. NFATc1 undergoes phosphorylation by NFATC-kinase and GSK3β and the phosphorylation induces its NFATc1 NuCLEAR exit. Its dephosphorylation by calcineurin promotes NuCLEAR import. The dephosphorylated form contains two unmasked NuCLEAR localization signals, which allow translocation of the protein to the nucleus. Ten different isoforms of NFATc1 have been described that are produced by alternative splicing and alternative initiation. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Serfling, E., et al. (2012). Cell Commun.  Sig. 10; Article 16; Chuvpilo, S.,  et al. (1999). J. Immunol. 162(12); 7294-7301; Beals, CR., et al. (1997). Science. 275(5308). 1930-1934).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
NuCLEAR is a trademark of Sigma-Aldrich Co. LLC
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service