Skip to Content
MilliporeSigma
All Photos(12)

Key Documents

ZRB2039

Sigma-Aldrich

Anti-C1QA Antibody, clone 1M12 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Complement C1q subcomponent subunit A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1M12, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 26.02 kDa
observed mol wt ~32 kDa

purified by

using Protein A

species reactivity

mouse, human, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... C1QA(712)

Related Categories

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1M12 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects C1QA. It targets an epitope within 22 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the internal region of human Complement C1q subcomponent subunit A (C1QA).

Application

Quality Control Testing

Evaluated by Western Blotting in PC12 cell lysates.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected C1QA in PC12 cell lysates.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected C1QA in lysates from Human kidney tissue and NIH3T3 cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected C1QA in HepG2 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected C1QA in human liver tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound C1QA peptide with a KD of 1.3 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Complement C1q subcomponent subunit A (UniProt: P02745; also known as C1QA) is encoded by the C1QA gene (Gene ID: 712) in human. C1q is the first subcomponent of the C1 complex of the classical pathway of complement activation. C1 complex circulates in plasma as an inactive  protein complex of one C1q subunit, two C1r subunits and two C1s subunits. This complex is formed by the calcium dependent interaction of C1q with C1r2C1s2. Initially, a conformational change in C1r occurs, followed by proteolytic activation, which results in the cleavage of all four polypeptide chains of C1r2C1s2. The two activated C1s subunits are then able to catalyze the assembly of the C3 convertase (C4b2a), which is formed from C2 and C4.  The C1q subcomponent of the complex is composed of nine subunits, six of which are disulfide-linked dimers of the A and B chains, and three of which are disulfide-linked dimers of the C chain. Its C1q domain is localized in amino acids 110-245. It also has a collagen-like domain (aa 31-109) that is involved in interaction with C1r2C1s2 proenzyme complex.  C1q plays an important recognition role in adaptive and innate immunity through its ability to interact, via its six globular head regions, with both immunoglobulin and non-immunoglobulin activators of the complement system. Mutations in C1QA gene have been linked to complement component C1q deficiency (C1QD) that leads to severe immune complex disease with features of systemic lupus erythematosus and glomerulonephritis. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Reid, KBM. ( 2018). Front. Immunol. 9; 764; Kishore, U., and Reid, KB (2000). Immunopharmacology. 49(1-2); 159-170).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service