Skip to Content
MilliporeSigma
All Photos(22)

Key Documents

ZRB2034

Sigma-Aldrich

Anti-Topoisomerase II Antibody, clone 3C3 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

DNA topoisomerase 2-alpha;EC:5.6.2.2 1;DNA topoisomerase II alpha isozyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ICC
IHC
WB
affinity binding assay
clone:
3C3, recombinant monoclonal
species reactivity:
mouse, human
citations:
technique(s):
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3C3, recombinant monoclonal

description

3C3 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 174.39 kDa
observed mol wt ~170 kDa

purified by

using Protein A

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TOP2A(7153)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3C3 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Topoisomerase II. It targets an epitope within 20 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of human Topoisomerase II.

Application

Quality Control Testing

Evaluated by Western Blotting in Neuro2a cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Topoisomerase II in Neuro2a cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Topoisomerase II in HeLa cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Topoisomerase II in human tonsil tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Topoisomerase II in A431, HeLa, HepG2, and NIH3T3 cells.

Affinity Binding Assay: A representative lot of this antibody bound Topoisomerase II peptide with a KD of 5.7 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

DNA topoisomerase 2-alpha (UniProt: P11388; also known as EC:5.6.2.2 1, DNA topoisomerase II, alpha isozyme) is encoded by the TOP2A (also known as TOP2) gene (Gene ID: 7153) in human. DNA topoisomerases are nuclear enzymes that regulate the conformational changes in DNA topology by catalyzing the breakage and rejoining of DNA strands during the normal cell cycle. They relieve torsional stress during replication and transcription. Three different types of topoisomerases have been reported in human. They are types 1; 2 , and 2β. They are encoded by genes on three separate chromosomes. Simpler organisms possess only topoisomerase 1; however, higher organisms have all three types of topoisomerases. Topoisomerase I reversibly cleaves a single strand in duplex DNA molecule, whereas topoisomerase 2 breaks and rejoins both DNA strands. Topoisomerase II is found at replication forks and remains tightly associated with chromosomes during mitosis. The N-terminus of Topoisomerase 2 contains several structural domains; the ATPase domain (about residues 1-265), the transducer domain (about 266-428) and the toprim domain (455-572). It is essential for the survival of actively growing cells. Its levels are upregulated dramatically during periods of cell proliferation. Its levels increase over the cell cycle and peak in G2/M phase. Topoisomerase II functions in growth-dependent processes, such as DNA replication and chromosome segregation. In contrast, the expression of the β isoform is independent of proliferative status and it dissociates from chromosomes during mitosis. Topoisomerase IIβ cannot compensate for the loss of topoisomerase II in mammalian cells. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: McClendon, AK., and Osheroff, N. (2007). Mutat. Res. 623(1-2); 83-97; Wyles, JP., et al. (2007). Nucleic Acids Res. 35(12); 4076-4085).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service