Skip to Content
MilliporeSigma
All Photos(13)

Key Documents

ZRB1987

Sigma-Aldrich

Anti-pan Rho (A/B/C) Antibody, clone 1I16 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Transforming protein RhoA;Rho-related GTP-binding protein RhoB;Rho-related GTP-binding protein RhoC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1I16, recombinant monoclonal

description

1I16 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 22.0 kDa
observed mol wt ~18-22 kDa

purified by

using Protein A

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1I16 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Rho A, B, and C. It targets an epitope within 22 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the internal region of human Rho A.

Application

Quality Control Testing

Evaluated by Western Blotting in HL-60 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Rho (A/B/C) in HL-60 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Rho (A/B/C) in lysates from Human brain tissue and Ramos, RAW264.7, and C6 cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Rho (A/B/C) in U2OS cells.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected Rho (A/B/C in one million U2OS cells.

Affinity Binding Assay: A representative lot of this antibody bound Rho A peptide with a KD of 4.2 x 10-8 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Rho (A/B/C) in human cerebral cortex tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Rho-related GTP binding proteins Rho A, B, and C (UniProt: P61586, P62745, and P08134, respectively) are encoded by the RHOA, RHOB, and RHOC genes (Gene ID: 387, 388, and 389, respectively) in human. Rho GTPases that belong to the Ras superfamily of small GTPases are molecular switches cycling between a GDP-bound off- and a GTP-bound on-state. Three Rho GTPases have been identified in higher vertebrates: RhoA, RhoB, and RhoC. They are lipid-anchored cell membrane proteins that are mainly localized in the cytoplasmic side of plasma membrane. Despite about 85% homology of sequence, they play distinct roles in cells. They are involved in cell differentiation, proliferation, apoptosis, and membrane trafficking. They regulate cytoskeletal organization and thus affect multiple cellular functions, including cell motility, polarity, and division. Their switch in states is tightly regulated by Rho GTPase Activating Proteins (RhoGAPs). They are widely reported to regulate actin organization and is shown to impact the motility of cancer cells and affect invasion and metastasis. They contain a highly conserved GTPase binding domain in the N-terminus that is responsible for GDP/GTP binding and GTP hydrolysis. Their C-terminus region consists of a geranylgeranyl group and carboxyl methylation extension. They are reported to contribute to carcinoma progression of the multiple cancer tissues. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Mosaddeghzadeh, N., and Ahmadian, MR. (2021). Cells.10(7); 1831; Wennerberg, K., et al. (2005) J. Cell Sci. 118(5); 843-846; Gudi, T., et al (2002). J. Biol. Chem. 277(40); 37382-37393; Ridley, AJ. (1997). Int. J. Biochem. Cell Biol. 29(11); 1225-1229).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service