Skip to Content
MilliporeSigma
All Photos(10)

Key Documents

ZRB1729

Sigma-Aldrich

Anti-CBP Antibody, clone 3N20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CREB-binding protein;EC: 2.3.1.48;Histone lysine acetyltransferase CREBBP;Protein-lysine acetyltransferase CREBBP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3N20, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 265.35 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

rat

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG

epitope sequence

N-terminus

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CREBBP(1387)

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3N20 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects CREB-binding protein (CBP). It targets an epitope within 25 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the N-terminal region of human CREB-binding protein (CBP).

Application

Quality Control Testing

Evaluated by Immunocytochemistry in MCF-7 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected CREB-binding protein (CBP) in MCF-7 cells.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound CREB-binding protein (CBP) peptide with a KD of 1.3 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CREB-binding protein (CBP) in human testis tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

CREB-binding protein (UniProt: Q92793; also known as EC: 2.3.1.48, Histone lysine acetyltransferase CREBBP, Protein-lysine acetyltransferase CREBBP) is encoded by the CREBBP (also known as CBP) gene (Gene ID: 1387) in human. CBP, a homolog of p300, serves as a transcriptional co-activator of various sequence-specific transcription factors that are involved in a wide array of cellular activities, such as DNA repair, cell growth, differentiation, and apoptosis. It possesses intrinsic acetyltransferase activity by which it acetylates histones, giving specific tag for transcription activation. It can also acetylate non-histone proteins, such as DExD-box helicase 21, interferon regulatory factor 2, and nuclear receptor coactivator 3. CBP facilitates the transcription of active genes by acting as adapter protein bridging DNA binding transcription factors to the basal transcription machinery and by loosening chromatin through histone tail acetylation. It binds specifically to phosphorylated CREB and enhances its transcriptional activity toward cAMP-responsive genes. CBP and p300 share overall 63% amino acid sequence identity and about 90% homology in their histone acetyltransferase domain (aa 1323-1700). Structurally, CBP contains several domains: a receptor-interacting domain (RID); KIX binding site of CREB; a bromo domain (aa 1103-1175); three cysteine and histidine-rich domains, a glutamine-and proline-rich domain (QP), and a steroid receptor co-activator-1 interaction domain (SID). Methylation of the KIX domain by coactivator associated arginine methyltransferase 1 (CARM1) blocks its association with CREB that results in the blockage of CREB signaling. Phosphorylation of CBP by IKK- at serine 1382 and 1386 promotes cell growth by switching the binding preference of CBP from TP53 to NF-kB. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Das, C., et al. (2014). Proc. Natl. Acad. Sci. USA. 111(12); E1072-E1081; Song, C., et al. (2017). Genes Dev. 31(13); 1370-1381; Karamouzis, MV., et al. (2007). Cell Res. 17(4); 324-332).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

0.3 mg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service