Skip to Content
MilliporeSigma
All Photos(12)

Key Documents

ZRB1684

Sigma-Aldrich

Anti-PML Antibody, clone 1F13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Protein PML;E3 SUMO-protein ligase PML;Promyelocytic leukemia protein;RING finger protein 71;RING-type E3 SUMO transferase PML;Tripartite motif-containing protein 19;TRIM19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F13, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 97.55 kDa
observed mol wt ~105 kDa

purified by

using Protein A

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PML(5371)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1F13 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Promyelocytic leukemia protein (PML). It targets an epitope within 19 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal region of human Promyelocytic leukemia protein (PML).

Application

Quality Control Testing

Evaluated by Western Blotting in NIH3T3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PML in NIH3T3 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PML in HeLa and L6 cell lysates.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected PML in A431 cells.

Affinity Binding Assay: A representative lot of this antibody bound PML with a KD of 1.1 x 10-8 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected PML in human colon tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Protein PML (UniProt: P29590; also known as E3 SUMO-protein ligase PML, Promyelocytic leukemia protein, RING finger protein 71, RING-type E3 SUMO transferase PML, Tripartite motif-containing protein 19, TRIM19) is encoded by the PML (also known as MYL, PP8675, RNF71, TRIM19) gene (Gene ID: 5371) in human. PML is the eponymous and main structural component of the PML nuclear bodies (PML NBs). It is involved in a variety of cellular processes, including tumor suppression, transcriptional regulation, apoptosis, senescence, DNA damage response, and viral defense mechanisms. It has been reported that PML recognizes misfolded proteins and acts as a SUMO E3 ligase by promoting the attachment of SUMO to these substrates. PML preferentially catalyzes modification with the SUMO2/SUMO3 variants that are prone to the formation of polymeric SUMO chains. SUMO2/SUMO3 chains subsequently serve as a docking site for RNF4, which earmarks the misfolded proteins for proteasomal degradation by adding a ubiquitin tag. PML protein is localized in the nucleus and in cytoplasm. 14 different isoforms of PML have been described that are produced via alternative splicing. Isoform PML-1 is shown to shuttle between the nucleus and cytoplasm and Isoforms 2-6 are localized in the nucleus. Some isoforms, such as PLML 7 and 14 lack nuclear localization signal and are only cytosolic. Acetylation at Lys-487 is shown to be essential for its nuclear localization. Within the nucleus, most of PML is expressed in the diffuse nuclear fraction of the nucleoplasm and only a small fraction is found in the matrix-associated PML-NBs. The transfer of PML from the nucleoplasm to PML-NBs is dependent on its phosphorylation and sumoylation. The B1 box (aa 124-166) and the RING finger (aa 57-92) zinc fingers are also required for the localization in PML-NBs. PML is also observed in mitochondria-associated membranes (MAMs) that connect the endoplasmic reticulum (ER) and the mitochondria. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Guo, L., et al. (2014). Mol. Cell 55(1); 15-30; Chu, Y., and Yang, X. (2011). Oncogene. 30(9): 1108-1116).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service