Skip to Content
MilliporeSigma
All Photos(9)

Documents

ZRB1681

Sigma-Aldrich

Anti-Calreticulin Antibody, clone 1F16 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CRP55

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F16, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 48.14 kDa
observed mol wt ~58 kDa

purified by

using Protein A

species reactivity

mouse, human

species reactivity (predicted by homology)

monkey, horse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CALR(811)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1F16 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Calreticulin. It targets an epitope within 21 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal half of human Calreticulin.

Application

Quality Control Testing

Evaluated by Western Blotting in U2OS cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Calreticulin in U2OS cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Calreticulin in NIH3T3 cell lysate.

Affinity Binding Assay: A representative lot of this antibody bound Calreticulin with a KD of 1.0 x 10-12 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Calreticulin in U2OS cells..

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Calreticulin (UniProt: P27797; also known as CRP55, Calregulin, Endoplasmic reticulum resident protein 60, ERp60, HACBP, grp60) is encoded by the CALR (also known as CRTC) gene (Gene ID: 811) in human. Calreticulin is a highly conserved, monomeric, endoplasmic reticulum protein that is synthesized with a signal peptide (aa 1-17), which is subsequently cleaved off to generate the mature form (aa 18-417). It serves as a calcium-binding chaperone that promotes folding, oligomeric assembly, and quality control in the endoplasmic reticulum via the calreticulin/calnexin cycle. It consists of three distinct domains: An N-terminal lectin-like globular domain that is required for its interaction with integrins and it contains a steroid receptor-like DNA-binding site that is involved in chaperone functions; a second domain is a central proline-rich domain that forms an elongated arm-like structure and has high-affinity and low-capacity for calcium-binding and participates in its chaperone activity. The third domain is a highly acidic, localized in the C-terminal region, displays low-affinity and high-capacity for Ca2+ binding and is involved in calcium -buffering functions of calreticulin. It also has a KDEL motif at the C-terminus that is responsible for calreticulin retrieval from the Golgi apparatus to the endoplasmic reticulum. Cancer cells undergoing immunogenic cell death are reported to expose calreticulin on their surface, which promotes the uptake of dead cells by phagocytes. Mutations leading to loss-of-function of CALR gene promote oncogenesis by impairing homeostasis in healthy cells and hamper natural and therapy-driven immunosurveillance. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Fucikova, J., et al. (2021). Cell Res. 31(1); 5-16; Krause, KH., and Michalak, M. (1997). Cell. 88(4); 439-443).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service