Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

ZRB1643

Sigma-Aldrich

Anti-Lubricin/PRG4 Antibody, clone 1C20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Proteoglycan 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C20, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 151.06 kDa
observed mol wt ~30 kDa

purified by

using Protein A

species reactivity

mouse, human

species reactivity (predicted by homology)

rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PRG4(10216)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C20 is a ZooMAb® Rabbit recombinant monoclonal antibody that detects Lubricin (Proteoglycan 4). It targets an epitope within 24 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the C-terminal region of human Lubricin (Proteoglycan 4).

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant human PRG4 protein.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected recombinant human Lubricin/PRG4.

Tested Applications


Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Lubricin/PRG4 in mouse cartilage tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound Lubricin/PRG4 with a KD of 1.6 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Proteoglycan 4 (UniProt: Q92954; also known as Lubricin, Megakaryocyte-stimulating factor, Superficial zone proteoglycan) is encoded by the PRG4 (also known as MSF, SZP) gene (Gene ID: 10216) in human. Proteoglycan 4 is a heavily O-glycosylated, secreted, mucin-like protein that is synthesized with a signal peptide (aa 1-24), which is subsequently cleaved off to generate the mature form (aa 25-1404) that can be further cleaved into a C-terminal part (aa 1307-1404). It is highly expressed in synovial tissue, cartilage, and liver and is weakly expressed in heart and lung. It is reported to play a role in boundary lubrication within articulating joints and prevents protein deposition onto cartilage from synovial fluid by controlling adhesion-dependent synovial growth and inhibiting the adhesion of synovial cells to the cartilage surface. Human lubricin contains two mucin-like domains. The first mucin domain contains multiple copies of the peptide motif KEPAPTTT, with threonine residues that are critical for O-linked GalNAc-Gal-NeuAc sidechains. The second mucin-like domain is also threonine rich but does not contain the KEPAPTTT motif. It also contains a hemopexin-like (PEX) domain at its C-terminal that is suggested to be essential for extracellular matrix binding. Lack of proteoglycan 4 expression leads to premature joint failure. Its low concentration in synovial fluid following an injury is associated with increased cartilage damage and may play a role in development of osteoarthritis. Proteoglycan 4 is reported to reduce neuroinflammation and protect the blood-brain barrier after traumatic brain injury. It restores blood-brain barrier function and prevents post-traumatic loss of tight junction protein claudin 5 and reduces neuronal death. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Bennett, M., et al. (2021). J. Neurotrauma. 38(4); 385-398; Iqbal, SM., et al. (2016). Sci. Rep. 6; Article 18910).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service