Skip to Content
MilliporeSigma
All Photos(13)

Key Documents

ZRB1620

Sigma-Aldrich

Anti-IFNγ Antibody, clone 5K15 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

IFN-gamma, Immune interferon, Interferon gamma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

clone

recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 19.35 kDa
observed mol wt ~15 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 5K15 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human Interferon- g.

Immunogen

Full-length mature recombinant human Interferon- g (without signal and propeptide).

Application

Quality Control Testing
Western Blotting Analysis (WB): Analysis: A 1:1,000 dilution of this antibody detected human IFN-g protein.Tested applications Western Blotting Analysis (WB): A 1:1000 Dilution of this antibody detected IFN- g in NK-92 cell lysate. Flow Cytometry Analysis: 1 μg from a representative lot detected IFN- in one million HuPBMC.Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected IFN- in NK-92 cells treated with Brefeldin A (BFA; 300 ng/ml, 4 hours) or stimmed and treated with BFA.Affinity Binding Assay:: A representative lot of this antibody bound IFN-g with a KD of 2.3 x 10-8 in an affinity binding assay.Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-IFN-g, clone 5K15 ZooMAb, Cat. No. ZRB1620, is a recombinant Rabbit monoclonal antibody that specifically targets IFN-g and is tested for use in Affinity Binding Assay, Flow Cytometry, Immunocytochemistry, and Western Blotting.

Target description

Interferon- g (UniProt: P01579; also known as IFN-gamma, IFN- g, Immune interferon) is encoded by the IFNG gene (Gene ID: 3458) in human. IFN-g is a pleiotropic molecule with antiproliferative, pro-apoptotic, and antitumor properties. It is a type II homodimeric interferon that is synthesized with a signal peptide (aa 1-23) and a propeptide (aa 162-166) that are subsequently cleaved off to produce the mature form. It is secreted predominantly by activated lymphocytes such as CD4 T helper type 1, CD8 cytotoxic T cells, g T cells, and natural killer cells, and antigen-presenting cells. Its expression is induced by mitogens and cytokines. IFN- g signaling coordinates several biological responses involved in host defense and immune surveillance, establishment of adaptive immunity, and the regulation of inflammation. Its diverse biological activities are initiated by IFN- g -mediated aggregation of at least two different cell surface receptors: IFN g R1 and IFN g R2. IFN-g is reported to activate macrophages toward a pro-inflammatory profile increasing their phagocytic ability and enhancing their microbial killing activity. It is also shown to upregulate cell surface MHC class II on antigen presenting cells, thus promoting peptide-specific activation of CD4 T cells. Mutations in IFNG gene are known to cause aplastic anemia that is characterized by failure of bone marrow to produce adequate number of peripheral blood elements leading to peripheral pancytopenia and hypoplasia. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Castro, F., et al. (2018). Front. Immunol. 9; 847).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service